Works great with heptamutant sortaseA-mediated ligations!
B9181
Biotin ethylenediamine hydrobromide
≥95% purity (TLC), solid
Synonym(s):
(3aS,4S,6aR)-N-(2-Aminoethyl)hexahydro- 2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide, N-(2-Aminoethyl)biotinamide hydrobromide
About This Item
Recommended Products
Product Name
Biotin ethylenediamine hydrobromide, ≥95% (TLC), solid
Quality Level
assay
≥95% (TLC)
form
solid
mol wt
367
color
white
solubility
DMF: >10 mg/mL
DMSO: >10 mg/mL
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-8°C
SMILES string
Br.NCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
InChI
1S/C12H22N4O2S.BrH/c13-5-6-14-10(17)4-2-1-3-9-11-8(7-19-9)15-12(18)16-11;/h8-9,11H,1-7,13H2,(H,14,17)(H2,15,16,18);1H/t8-,9-,11-;/m0./s1
InChI key
SGNZEZHLDUVMMU-GRIHUTHFSA-N
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service