Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4630

Sigma-Aldrich

Ammonium sulfamate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SO3NH2
CAS Number:
Molecular Weight:
114.12
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

≥99%

mp

131-135 °C (lit.)

SMILES string

N.NS(O)(=O)=O

InChI

1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3

InChI key

GEHMBYLTCISYNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service