Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86962

Sigma-Aldrich

1,1,2,2-Tetrachloroethane

purum, ≥95.0% (GC)

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein/REAXYS Number:
969206
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

grade

purum

assay

≥95.0% (GC)

color

slightly green-yellow

refractive index

n20/D 1.494 (lit.)
n20/D 1.494

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformations of 1, 1, 2, 2-tetrachloroethane under methanogenic conditions.
Chen C, et al.
Environmental Science & Technology, 30(2), 542-547 (1996)
1, 1, 2, 2-Tetrachloroethane (TeCA) as a Solvent Additive for Organic Hole Transport Materials and Its Application in Highly Efficient Solid-State Dye-Sensitized Solar Cells.
Xu B, et al.
Advanced Energy Materials, 5(10) (2015)
Srigokul Upadhyayula et al.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
Debolina Basu et al.
Bioresource technology, 101(1), 21-25 (2009-08-25)
This study examines the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor in treating simulated wastewaters containing 1,1,2,2-tetrachloroethane (TeCA). Reactors R1 (control) and R2 (containing TeCA) were operated at Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and
Wan-Chun Tsai et al.
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service