Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

858404

Sigma-Aldrich

Methyl violet 2B

indicator grade, Dye content 75 %

Synonym(s):

Basic Violet 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
42535
Beilstein/REAXYS Number:
8464808
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

indicator grade

composition

Dye content, 75%

mp

137 °C (dec.) (lit.)

λmax

584 nm

SMILES string

Cl.C\N=C1/C=C\C(C=C1)=C(/c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

InChI

1S/C24H27N3.ClH/c1-25-21-12-6-18(7-13-21)24(19-8-14-22(15-9-19)26(2)3)20-10-16-23(17-11-20)27(4)5;/h6-17H,1-5H3;1H

InChI key

JFTBTTPUYRGXDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Violet 2B is a mixture of polymethylated (4,5 and 6) pararosaniline chlorides

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service