Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69880

Sigma-Aldrich

Morpholine

puriss. p.a., ACS reagent, ≥99.0% (GC)

Synonym(s):

Tetrahydro-1,4-oxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
Beilstein/REAXYS Number:
102549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

vapor density

3 (vs air)

vapor pressure

31 mmHg ( 38 °C)
7 mmHg ( 20 °C)

assay

≥99.0% (GC)

autoignition temp.

590 °F

expl. lim.

10.8 %

impurities

≤0.3% water

refractive index

n20/D 1.454 (lit.)
n20/D 1.455

bp

129 °C (lit.)
129 °C

mp

−7-−5 °C (lit.)

density

1.000 g/mL at 20 °C
0.996 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1COCCN1

InChI

1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2

InChI key

YNAVUWVOSKDBBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

87.8 °F - closed cup

flash_point_c

31 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammednoor Altarawneh et al.
The journal of physical chemistry. A, 116(29), 7703-7711 (2012-07-04)
The combustion chemistry of morpholine (C(4)H(8)ONH) has been experimentally investigated recently as a representative model compound for O- and N-containing structural entities in biomass. Detailed profiles of species indicate the self-breakdown reactions prevailing over oxidative decomposition reactions. In this study
Sean E Low et al.
Journal of neurophysiology, 108(1), 148-159 (2012-04-12)
The molecular and physiological basis of the touch-unresponsive zebrafish mutant fakir has remained elusive. Here we report that the fakir phenotype is caused by a missense mutation in the gene encoding voltage-gated calcium channel 2.1b (CACNA1Ab). Injection of RNA encoding
Su-Juan Li et al.
ACS nano, 4(11), 6417-6424 (2010-10-21)
A strategy for label-free oligonucleotide (DNA) analysis has been proposed by measuring the DNA-morpholino hybridization hindered diffusion flux of probe ions Fe(CN)(6)(3-) through nanochannels of a porous anodic alumina (PAA) membrane. The flux of Fe(CN)(6)(3-) passing through the PAA nanochannels
M F Kasakin et al.
Bioorganicheskaia khimiia, 37(6), 830-835 (2012-04-14)
A simple and effective method for the synthesis of 2'-aminomethylmorpholino-4'-carboxymethyl nucleoside analogues and Boc-modified derivatives as synthons for peptide synthesis was developed.
Nickel-catalyzed amination of aryl sulfamates.
Stephen D Ramgren et al.
Angewandte Chemie (International ed. in English), 50(9), 2171-2173 (2011-02-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service