67994
N-Ethyldiisopropylamine
produced by BASF, ≥98.0% (GC)
Synonym(s):
N,N-Diisopropylethylamine, Hünigs base
About This Item
Recommended Products
grade
produced by BASF
assay
≥98.0% (GC)
bp
126-128 °C (lit.)
density
0.755 g/mL at 20 °C (lit.)
0.757 g/mL at 25 °C (lit.)
SMILES string
CCN(C(C)C)C(C)C
InChI
1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3
InChI key
JGFZNNIVVJXRND-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
49.1 °F
flash_point_c
9.5 °C
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service