Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66750

Sigma-Aldrich

Dichloromethane

purum, ≥98.0% (GC)

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein/REAXYS Number:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

purum

assay

≥98.0% (GC)

autoignition temp.

1223 °F

contains

~0.002% amylene as stabilizer

expl. lim.

22 %

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Sales restrictions may apply

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic peptide synthesis in organic solvent with different zeolites as immobilization matrixes.
Xing GW, et al.
Tetrahedron, 56(22), 3517-3522 (2000)
R Re et al.
Free radical biology & medicine, 26(9-10), 1231-1237 (1999-06-25)
A method for the screening of antioxidant activity is reported as a decolorization assay applicable to both lipophilic and hydrophilic antioxidants, including flavonoids, hydroxycinnamates, carotenoids, and plasma antioxidants. The pre-formed radical monocation of 2,2'-azinobis-(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS*+) is generated by oxidation
V Melissinaki et al.
Biofabrication, 3(4), 045005-045005 (2011-09-21)
This study reports on the production of high-resolution 3D structures of polylactide-based materials via multi-photon polymerization and explores their use as neural tissue engineering scaffolds. To achieve this, a liquid polylactide resin was synthesized in house and rendered photocurable via
Effect of activated carbon content in TiO2-loaded activated carbon on photodegradation behaviors of dichloromethane.
Torimoto T, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 103(1), 153-157 (1997)
Dipyridine-chromium (VI) oxide oxidation of alcohols in dichloromethane.
Collins JC, et al.
Tetrahedron Letters, 9(30), 3363-3366 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service