Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

639133

Supelco

Acetonitrile solution

suitable for HPLC, contains 0.1 % (v/v) ammonium hydroxide

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein/REAXYS Number:
4124158
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
technique(s):
HPLC: suitable
bp:
81-82 °C
vapor pressure:
72.8 mmHg

Product Name

Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide

vapor density

1.41 (vs air)

Quality Level

vapor pressure

72.8 mmHg

form

liquid

contains

0.1 % (v/v) ammonium hydroxide

expl. lim.

8 %

technique(s)

HPLC: suitable

bp

81-82 °C

mp

-48 °C

density

0.782 g/mL at 25 °C

format

mixture

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is an acetonitrile solution containing 0.1%v/v ammonium hydroxide that can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. This pre-blended solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. Acetonitrile (MeCN), an organic solvent[1] has low viscosity, high elution strength and is UV transparent at low wavelength.[2]

Preparation Note

Product filtered through a 0.2 μm filter

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)
Acetonitrile, the polarity chameleon.
Zarzycki PK, et al.
Analytical and Bioanalytical Chemistry, 397(3), 905-908 (2010)
Kangmin Chon et al.
Water research, 81, 388-397 (2015-07-05)
In this study, the changes in UV absorbance at 254 nm (UVA254) and electron donating capacity (EDC) were investigated as surrogate indicators for assessing removal of micropollutants and bromate formation during ozonation of wastewater effluents. To measure the EDC, a novel
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations
Maria de la Paz Celorio-Mancera et al.
Insect biochemistry and molecular biology, 61, 34-45 (2015-05-03)
More than half of the proteome from mandibular glands in caterpillars is represented by chemosensory proteins. Based on sequence similarity, these proteins are putative transporters of ligands to gustatory receptors in sensory organs of insects. We sought to determine whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service