Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

59309

Sigma-Aldrich

2-Propanol

puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.5% (GC)

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein/REAXYS Number:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

grade

absolute
puriss.

assay

≥99.5% (GC)

autoignition temp.

750 °F

quality

over molecular sieve (H2O ≤0.005%)

expl. lim.

2.0-12.7 %, 93 °C

refractive index

n20/D 1.377 (lit.)
n20/D 1.377

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of three different TiO 2 morphologies on hydrogen evolution by methanol assisted water splitting: nanoparticles, nanotubes and aerogels
D?Elia D, et al.
International Journal of Hydrogen Energy, 36(22), 14360-14373 (2011)
A direct 2-propanol polymer electrolyte fuel cell.
Cao D and Bergens SH.
Journal of Power Sources, 124(1), 12-17 (2003)
Electrochemical photocatalytic degradation of dye solution with a TiO2-coated stainless steel electrode prepared by electrophoretic deposition.
Lin WC, et al.
Applied Catalysis. B, Environmental, 140, 32-41 (2013)
Formation and characteristics of Cu colloids from CuO powder by laser irradiation in 2-propanol.
Yeh MS, et al.
The Journal of Physical Chemistry B, 103(33), 6851-6857 (1999)
Evaluation of Ethanol, 1-Propanol, and 2-Propanol in a Direct Oxidation Polymer-Electrolyte Fuel Cell A Real-Time Mass Spectrometry Study.
Wang J, et al.
Journal of the Electrochemical Society, 142(12), Wang J-Wang J (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service