554049
Tetrazole solution
reagent grade, 3 wt. % in acetonitrile
About This Item
Recommended Products
grade
reagent grade
concentration
3 wt. % in acetonitrile
impurities
≤20 ppm water
bp
84 °C
mp
156-158 °C (lit.)
SMILES string
c1nnn[nH]1
InChI
1S/CH2N4/c1-2-4-5-3-1/h1H,(H,2,3,4,5)
InChI key
KJUGUADJHNHALS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
41.0 °F - closed cup
flash_point_c
5 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service