Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539325

Sigma-Aldrich

Hydrofluoric acid

35 wt. % in H2O

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
EC Number:
MDL number:
UNSPSC Code:
12352106

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

concentration

35 wt. % in H2O

density

1.15 g/mL at 25 °C (lit.)
1.15 g/mL at 25 °C

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Souleymane Coulibaly et al.
Biological & pharmaceutical bulletin, 38(9), 1395-1403 (2015-09-04)
To clarify the seasonal fluctuations in air pollution and the effect of long-range transport, we collected airborne particles (n=118) at Dazaifu in Fukuoka, Japan, from June 2012 to May 2013 and measured Pb and SO4(2-), which are indicators of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service