Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510912

Sigma-Aldrich

Sodium nitrite

powder, −325 mesh, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

97%

form

powder

autoignition temp.

914 °F

particle size

−325 mesh

mp

271 °C (lit.)

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiffany A Major et al.
Antimicrobial agents and chemotherapy, 54(11), 4671-4677 (2010-08-11)
A hallmark of airways in patients with cystic fibrosis (CF) is highly refractory, chronic infections by several opportunistic bacterial pathogens. A recent study demonstrated that acidified sodium nitrite (A-NO(2)(-)) killed the highly refractory mucoid form of Pseudomonas aeruginosa, a pathogen
Paul J Shami et al.
Journal of medicinal chemistry, 49(14), 4356-4366 (2006-07-11)
The literature provides evidence that metabolic nitric oxide (NO) release mediates the cytotoxic activities (against human leukemia and prostate cancer xenografts in mice) of JS-K, a compound of structure R(2)N-N(O)=NO-Ar for which R(2)N is 4-(ethoxycarbonyl)piperazin-1-yl and Ar is 2,4-dinitrophenyl. Here
Deniz Ekinci et al.
Bioorganic & medicinal chemistry, 19(2), 744-748 (2011-01-08)
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of
Alessandra Cincinelli et al.
Bioorganic & medicinal chemistry, 19(6), 1847-1851 (2011-03-08)
A high activity α-carbonic anhydrase (CA, EC 4.2.1.1) has been purified from various tissues of the Antarctic seal Leptonychotes weddellii. The new enzyme, denominated lwCA, has a catalytic activity for the physiologic CO(2) hydration to bicarbonate reaction, similar to that
Peter Burghout et al.
Bioorganic & medicinal chemistry, 19(1), 243-248 (2010-12-18)
The Gram-positive bacterium Streptococcus pneumoniae is a human respiratory tract pathogen that contributes significantly to global mortality and morbidity. It was recently shown that this bacterial pathogen depends on a conserved β-carbonic anhydrase (CA, EC 4.2.1.1) for in vitro growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service