Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47590

Sigma-Aldrich

Hydrofluoric acid

puriss. p.a., ≥40%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
MDL number:
UNSPSC Code:
12352106

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

grade

puriss. p.a.

assay

≥40% (acidimetric)
≥40%

ign. residue

≤0.0005% (as SO4)

density

1.15 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
hexafluorosilicate (SiF62-): ≤50 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤2 mg/kg
sulfite (SO32-): ≤2 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.05 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.2 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.02 mg/kg
Mg: ≤0.2 mg/kg
Mn: ≤0.05 mg/kg
Mo: ≤0.05 mg/kg
Na: ≤0.2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.02 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

in plastic bottles

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Reagent for the decomposition of silicates[1][2]

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
Denise C Jarrin et al.
Pediatric cardiology, 36(1), 41-48 (2014-07-25)
Heart rate variability (HRV) is a non-invasive quantitative marker of cardiac autonomic function derived from continuous electrocardiogram (ECG) recordings. Normative HRV values and development factors have not been established in pediatric populations. The objective was to derive referent time- and
Nicolas Vodovar et al.
JACC. Heart failure, 3(8), 629-636 (2015-08-08)
This study sought to hypothesize that elevated B-type natriuretic peptide (BNP) could act as an endogenous neprilysin inhibitor. A hallmark of acute decompensated heart failure (ADHF) is the overproduction of natriuretic peptides (NPs) by stretched cardiomyocytes. Various strategies have been
Philip J Millar et al.
Circulation, 131(5), 459-468 (2014-12-04)
Muscle sympathetic activation in heart failure with reduced ejection fraction (HFrEF) has been attributed, on the basis of multiunit recordings, to attenuated inhibitory feedback from stretch-sensitive cardiopulmonary mechanoreceptors. However, such preparations integrate 2 populations of single units exhibiting directionally opposite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service