Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46002

Sigma-Aldrich

4-Methylumbelliferyl acetate

for fluorescence

Synonym(s):

MU-Ac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O4
CAS Number:
Molecular Weight:
218.21
Beilstein/REAXYS Number:
189667
EC Number:
MDL number:
UNSPSC Code:
12352204

grade

for fluorescence

mp

149-150 °C (lit.)

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 365 nm; λem 445 nm in 0.1 M Tris pH 8.0 (esterase)

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C12H10O4/c1-7-5-12(14)16-11-6-9(15-8(2)13)3-4-10(7)11/h3-6H,1-2H3

InChI key

HXVZGASCDAGAPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Fluorogenic substrate for esterases[1]; Measurement of intracellular pH in rat proximal convoluted tubule[2]

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Dive et al.
Cytometry, 8(6), 552-561 (1987-11-01)
Flow cytoenzymology is the determination of enzyme activities or concentrations in single intact cells. Using the flow cytometer built and designed in our laboratory and recent modifications to hardware and software, we have developed an improved dynamic flow cytoenzymological procedure
E Pastoriza-Munoz et al.
The Journal of clinical investigation, 80(1), 207-215 (1987-07-01)
In the proximal convoluted tubule (PT), the HCO3- reabsorptive rate is higher in early (EPS) compared with late proximal segments (LPS). To examine the mechanism of this HCO3- reabsorption profile, intracellular pH (pHi) was measured along the superficial PT of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service