Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45330

Sigma-Aldrich

(±)-Epichlorohydrin

puriss., ≥99.5% (GC)

Synonym(s):

(±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
Beilstein/REAXYS Number:
79785
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.2 (vs air)

Quality Level

vapor pressure

13.8 mmHg ( 21.1 °C)

grade

puriss.

assay

≥99.5% (GC)

autoignition temp.

779 °F

expl. lim.

21 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.06% water

refractive index

n20/D 1.438 (lit.)
n20/D 1.438

bp

115-117 °C (lit.)

mp

−57 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

ClCC1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

BRLQWZUYTZBJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service