Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

419001

Sigma-Aldrich

Azure B thiocyanate

Dye content 98 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N4S2
CAS Number:
Molecular Weight:
328.46
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 98%

mp

228 °C (dec.) (lit.)

λmax

638 nm

SMILES string

[N-]=C=S.CNc1ccc2N=C3C=CC(\C=C3Sc2c1)=[N+](/C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service