Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

41648

Sigma-Aldrich

Dimethyl sulfoxide

puriss., absolute, over molecular sieve (H2O ≤0.01%), ≥99.5% (GC)

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

grade

absolute
puriss.

assay

≥99.5% (GC)

autoignition temp.

573 °F

quality

over molecular sieve (H2O ≤0.01%)

expl. lim.

42 %, 63 °F

refractive index

n20/D 1.479 (lit.)
n20/D 1.479

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parallel Protein Detection by Solid-Phase Proximity Ligation Assay with Real-Time PCR or Sequencing.
Ebai T, et al.
Current Protocols in Molecular Biology, 20-10 (2015)
Vincent Gache et al.
Methods in molecular medicine, 137, 29-43 (2007-12-19)
Microtubule-binding proteins are conveniently divided into two large groups: MAPs (microtubule-associated proteins), which can stabilize, anchor, and/or nucleate microtubules, and motors, which use the energy of ATP hydrolysis for a variety of functions, including microtubule network organization and cargo transportation
Oxidation of alcohols by ?activated? dimethyl sulfoxide. A preparative, steric and mechanistic study.
Omura K and Swern D.
Tetrahedron, 34(11), 1651-1660 (1978)
A RAPID PERMETHYLATION OF GLYCOLIPID, AND POLYSACCHARIDE CATALYZED BY METHYLSULFINYL CARBANION IN DIMETHYL SULFOXIDE.
S HAKOMORI
Journal of biochemistry, 55, 205-208 (1964-02-01)
Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service