402877
1,1,1-Trichloroethane
ACS reagent, ≥99%, contains 0.05% low alkyl epoxide as stabilizer
Synonym(s):
Methylchloroform, ‘Chlorothene’
About This Item
Recommended Products
grade
ACS reagent
vapor density
4.6 (vs air)
vapor pressure
100 mmHg ( 20 °C)
assay
≥99%
98.5-101.0% CH3CCl3 + active additives basis
autoignition temp.
998 °F
contains
0.05% low alkyl epoxide as stabilizer
expl. lim.
7.5-15 %, 25 °F
impurities
≤0.0003 meq/g Titr. acid
≤0.10% water
evapn. residue
≤0.001%
color
APHA: ≤10
refractive index
n20/D 1.4366 (lit.)
bp
74-76 °C (lit.)
mp
−35 °C (lit.)
density
1.336 g/mL at 20 °C (lit.)
SMILES string
CC(Cl)(Cl)Cl
InChI
1S/C2H3Cl3/c1-2(3,4)5/h1H3
InChI key
UOCLXMDMGBRAIB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ozone 1 - Skin Irrit. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service