40248
N,N-Dimethylformamide
puriss., absolute, over molecular sieve (H2O ≤0.01%), ≥99.8% (GC)
Synonym(s):
DMF, NSC 5356
About This Item
Recommended Products
vapor density
2.5 (vs air)
vapor pressure
2.7 mmHg ( 20 °C)
grade
absolute
puriss.
assay
≥99.8% (GC)
autoignition temp.
833 °F
quality
over molecular sieve (H2O ≤0.01%)
expl. lim.
15.2 %
impurities
≤0.005% free alkali (as NH3)
refractive index
n20/D 1.430 (lit.)
n20/D 1.430
bp
153 °C (lit.)
mp
−61 °C (lit.)
density
0.944 g/mL (lit.)
SMILES string
[H]C(=O)N(C)C
InChI
1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3
InChI key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
135.5 °F - closed cup
flash_point_c
57.5 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service