393517
Potassium hexacyanoferrate(III)
ACS reagent, ≥99%, powder, <10 μm
Synonym(s):
Potassium ferricyanide, Red prussiate
About This Item
grade
ACS reagent
assay
≥99%
form
powder
reaction suitability
reagent type: catalyst
core: iron
impurities
≤0.005% insolubles
≤0.05% ferro compounds ( as [Fe(CN)6]4- )
particle size
<10 μm
anion traces
chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%
SMILES string
[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N
InChI
1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1
InChI key
MIMJFNVDBPUTPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2
supp_hazards
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service