Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38070

Sigma-Aldrich

1,8-Diiodooctane

purum, ≥97.0% (GC)

Synonym(s):

Octamethylene diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)8I
CAS Number:
Molecular Weight:
366.02
Beilstein/REAXYS Number:
1735437
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.564
n20/D 1.5653 (lit.)

bp

167-169 °C/6 mmHg (lit.)

density

1.84 g/mL at 25 °C (lit.)

SMILES string

ICCCCCCCCI

InChI

1S/C8H16I2/c9-7-5-3-1-2-4-6-8-10/h1-8H2

InChI key

KZDTZHQLABJVLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service