Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376604

Sigma-Aldrich

4′,5′-Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein

Dye content 75 %

Synonym(s):

Calcein, DCAF, Fluorescein-4′,5′-bis(methyliminodiacetic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26N2O13
CAS Number:
Molecular Weight:
622.53
UNSPSC Code:
12171500

composition

Dye content, 75%

λmax

442 nm

SMILES string

OC(=O)CN(CC(O)=O)Cc1c(O)ccc2c1Oc3c(CN(CC(O)=O)CC(O)=O)c(O)ccc3C24OC(=O)c5ccccc45


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service