Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34932

Sigma-Aldrich

1,1,2-Trichloro-1,2,2-trifluoroethane

≥99.9% (GC)

Synonym(s):

1,1,2-Trichloro-trifluoroethane, Genetron® 113

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2CCl2F
CAS Number:
Molecular Weight:
187.38
Beilstein/REAXYS Number:
1740335
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor pressure

5.5 psi ( 20 °C)

assay

≥99.9% (GC)

autoignition temp.

1256 °F

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as CH3COOH)
≤0.005% water (Karl Fischer)

transmittance

235 nm, ≥25%
250 nm, ≥85%
265 nm, ≥98%

refractive index

n20/D 1.358 (lit.)

bp

47-48 °C (lit.)

mp

−35 °C (lit.)

density

1.57 g/mL at 25 °C (lit.)

suitability

passes test for IR spectroscopy
passes test for NMR

storage temp.

2-8°C

SMILES string

FC(F)(Cl)C(F)(Cl)Cl

InChI

1S/C2Cl3F3/c3-1(4,6)2(5,7)8

InChI key

AJDIZQLSFPQPEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Legal Information

Genetron is a registered trademark of Honeywell International Inc.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Ozone 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

383.0 °F

flash_point_c

195 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mette Christophersen et al.
Journal of contaminant hydrology, 81(1-4), 1-33 (2005-08-17)
An emplaced hydrocarbon source field experiment was conducted in the relatively homogeneous sandy geology of the vadose zone at Airbase Vaerløse, Denmark. The source (10.2 l of NAPL) consisted of 13 hydrocarbons (n-, iso- and cyclo-alkanes and aromates) and CFC-113
Y Michael et al.
International journal of pharmaceutics, 221(1-2), 165-174 (2001-06-09)
The nature of the drug-drug aggregation phenomena between salmeterol xinafoate and fluticasone propionate used in a metered-dose inhaler system has been examined. Interactions between the drugs in the solvents 1,1,2-trichlorotrifloroethane (CFC-113) and 1,1,1,2-tetrafluoroethane (HFA-134a) have been characterised using a focused
J A Richt et al.
Medical microbiology and immunology, 182(5), 271-280 (1993-11-01)
Treatment of homogenates from Borna disease virus (BDV)-infected brain tissue or cell cultures with Freon-113 yielded infectious particles with a buoyant density of 1.16-1.22 g/ml. Positive- and negative-stranded BDV-specific RNA species as well as three virus-specific proteins, known to be
V M Voge
Southern medical journal, 89(5), 516-518 (1996-05-01)
Freons are generally considered to be minimally toxic. There are no reports in the literature of Freons causing secondary arterial hypertension. We report two cases of acute, massive Freon exposure that preceded secondary arterial hypertension. We hypothesize that the arterial
H K Xiao et al.
American Industrial Hygiene Association journal, 51(7), 402-404 (1990-07-01)
Results obtained using Fourier transform infrared spectrophotometry (FTIR) for the analysis of samples of carbon disulfide (CS2) eluates containing trichloroethylene (TCE) and Freon from charcoal air sampling tubes were evaluated by comparison with results obtained when using gas chromatography (GC).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service