All Photos(1)
About This Item
Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352300
vapor pressure
<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)
mp
734 °C (lit.)
SMILES string
[K+].[Br-]
InChI
1S/BrH.K/h1H;/q;+1/p-1
InChI key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
May be used for removal of peripheral membrane proteins.
Not intended for use with FT-IR spectrometers
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zai-Chao Ma et al.
Marine biotechnology (New York, N.Y.), 17(4), 511-522 (2015-05-20)
In this study, in order to directly and efficiently convert inulin into pullulan, the INU1 gene from Kluyveromyces maximum KM was integrated into the genomic DNA and actively expressed in the high pullulan producer Aureobasidium melanogenum P16 isolated from the
Hao-Long Zhou et al.
Nature communications, 6, 6917-6917 (2015-04-22)
The dynamic behaviours of host frameworks and guest molecules have received much attention for their great relevance with smart materials, but little has been developed to control or understand the host-guest interplay. Here we show that the confined guest can
Igor L Dalinger et al.
Chemistry, an Asian journal, 10(9), 1987-1996 (2015-06-23)
A new family of energetic compounds, nitropyrazoles bearing a trinitromethyl moiety at the nitrogen atom of the heterocycle, was designed. The desirable high-energy dense oxidizers 3,4-dinitro- and 3,5-dinitro-1-(trinitromethyl)pyrazoles were synthesized in good yields by destructive nitration of the corresponding 1-acetonylpyrazoles.
Sophie Rodrigues et al.
Probiotics and antimicrobial proteins, 7(1), 45-51 (2014-10-22)
Vibrio tapetis CECT4600 is a pathogenic Gram-negative bacterium causing the brown ring disease in the Manila clam Ruditapes philippinarum. This vibriosis is induced by bacterial attachment on the periostracal lamina, yielding a decalcification of the bivalve shell. As in many
Stephanie A DeJong et al.
Applied spectroscopy, 69(6), 733-748 (2015-05-09)
Derivatives are common preprocessing tools, typically implemented as Savitzky-Golay (SG) smoothing derivatives. This work discusses the implementation and optimization of fourth-order gap derivatives (GDs) as an alternative to SG derivatives for processing infrared spectra before multivariate calibration. Gap derivatives approximate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service