Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337978

Sigma-Aldrich

Hydrogen bromide solution

30 wt. % in propionic acid

Synonym(s):

HBr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
Molecular Weight:
80.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

concentration

30 wt. % in propionic acid

density

1.311 g/mL at 25 °C

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service