323551
Sulfuric acid, fuming
ACS reagent, 20% free SO3 basis
Synonym(s):
Oleum
About This Item
Recommended Products
grade
ACS reagent
vapor density
3 (vs air)
vapor pressure
2 mmHg ( 25 °C)
concentration
20% (free SO3)
ign. residue
≤0.002%
color
colorless to very light brown
density
1.925 g/mL at 25 °C (lit.)
anion traces
nitrate (NO3-): ≤1 ppm
cation traces
As: ≤0.03 ppm
Fe: ≤2 ppm
NH4+: ≤3 ppm
SMILES string
O=S(=O)=O.OS(O)(=O)=O
InChI
1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);
Inchi Key
HIFJUMGIHIZEPX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service