31413
Magnesium chloride hexahydrate
puriss. p.a., ACS reagent, reag. ISO, ≥99%
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
agency
reag. ISO
assay
≥99%
impurities
≤0.0005% heavy metals (as Pb)
≤0.001% ammonium (NH4)
≤0.005% in water insoluble matters
pH
5-6.5 (20 °C, 5%)
anion traces
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg
cation traces
Ba: ≤20 mg/kg
Ca: ≤100 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤10 mg/kg
Sr: ≤20 mg/kg
Zn: ≤10 mg/kg
SMILES string
O.O.O.O.O.O.Cl[Mg]Cl
InChI
1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
InChI key
DHRRIBDTHFBPNG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service