Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31402

Sigma-Aldrich

Ammonium molybdate tetrahydrate

puriss. p.a., reag. Ph. Eur., ≥99%

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

agency

reag. Ph. Eur.

grade

puriss. p.a.

assay

≥99%

impurities

≤0.001% heavy metals (as Pb)

density

2.498 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤30 mg/kg
phosphate, arsenate, silicate (as SiO2): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤5 mg/kg

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Ho Park et al.
Journal of hazardous materials, 138(2), 311-316 (2006-07-25)
The petroleum refining industry makes extensive use of hydroprocessing catalysts. These catalysts contain environmentally critical and economically valuable metals such as Mo, V, Ni and Co. In the present study, a simple hydrometallurgical processing of spent hydrodesulphurization (HDS) catalyst for
Sidney Omelon et al.
Electrophoresis, 28(16), 2808-2811 (2007-07-21)
Nonradioactive polyphosphate (poly(P); (PO(3) (-))(n)) species resolved by PAGE can be detected by hydrolytic degradation of the polyphosphates into orthophosphates (P(i)) with a 5 M HCl solution saturated with NaCl, followed by staining the P(i) degradation products in a 1
Pradeep Verma et al.
Bioresource technology, 102(4), 3941-3945 (2010-12-24)
Pretreatments for enzymatic saccharification are crucial for the establishment of lignocellulosic biorefineries. In this study, we focused on ammonium ions and peroxometal complexes as potential delignifying agents. We first examined the pretreatment of beech wood with nine different ammonium salts
Shifeng Cao et al.
International journal of food microbiology, 141(3), 173-176 (2010-07-08)
The potential enhancement of Pichia membranifaciens by ammonium molybdate (NH(4)Mo) to control blue mould caused by Penicillium expansum on peach fruit was investigated. Combining P. membranifaciens at 1x10(8) cell/ml with 1 mM NH(4)Mo provided a more effective control of blue
W Grünberg et al.
Journal of dairy science, 98(8), 5385-5400 (2015-06-01)
Hypophosphatemia is a common finding in periparturient and anorectic cattle. Although the clinical relevance of hypophosphatemia in cattle is uncertain, it has been empirically associated with persistent recumbency, specifically in periparturient dairy cows. The objective of the present study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service