Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31274

Supelco

3,4-Dichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein/REAXYS Number:
1907693
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

~253 °C (lit.)

mp

65-67 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccc(Cl)c(Cl)c1

InChI

1S/C6H4Cl2O/c7-5-2-1-4(9)3-6(5)8/h1-3,9H

InChI key

WDNBURPWRNALGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Carey et al.
Canadian journal of physiology and pharmacology, 62(8), 971-975 (1984-08-01)
The disappearance rates of 2,4- and 3,4-dichlorophenol in a small stream were studied and were shown to be first order with respect to either distance or time of flow. Both chlorophenols disappeared at approximately the same rate with average half-lives
Julien Michel et al.
Journal of the American Chemical Society, 131(18), 6356-6357 (2009-05-07)
There is great interest in molecules capable of inhibiting the interactions between p53 and its negative regulators hDM2 and hDMX, as these molecules have validated potential against cancers in which one or both oncoproteins are overexpressed. We reported previously that
F O Bryant et al.
Applied and environmental microbiology, 57(8), 2293-2301 (1991-08-01)
The reductive dechlorination of pentachlorophenol (PCP) was investigated in anaerobic sediments that contained nonadapted or 2,4- or 3,4-dichlorophenol (DCP)-adapted microbial communities. Adaptation of sediment communities increased the rate of conversion of 2,4- or 3,4-DCP to monochlorophenols (CPs) and eliminated the
R Duran et al.
Applied microbiology and biotechnology, 59(2-3), 284-288 (2002-07-12)
Extracellular peroxidases play an important role in the degradation of chlorophenols by Phanerochaete chrysosporium. Depending on the moment of 3,4-dichlorophenol addition, the production of lignin peroxidase and manganese peroxidase in C-limited agitated cultures was affected in opposite ways. In cultures
P Larsson et al.
Applied and environmental microbiology, 54(7), 1864-1867 (1988-07-01)
The microbial degradation of a number of 14C-labeled, recalcitrant, aromatic pollutants, including trichloroguaiacol and di-, tri-, and pentachlorophenol, was investigated in aquatic model systems in the laboratory. Natural, mixed cultures of microorganisms in the water from a brown-water lake with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service