Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

287687

Sigma-Aldrich

Cyclopentane

anhydrous, ≥75% cyclopentane basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
Beilstein/REAXYS Number:
1900195
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

anhydrous

vapor density

~2 (vs air)

vapor pressure

18.93 psi ( 55 °C)
5.67 psi ( 20 °C)

assay

≥75% cyclopentane basis

autoignition temp.

682 °F

expl. lim.

8.7 %

evapn. residue

<0.0003%

refractive index

n20/D 1.405 (lit.)

bp

50 °C (lit.)

mp

−94 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

SMILES string

C1CCCC1

InChI

1S/C5H10/c1-2-4-5-3-1/h1-5H2

InChI key

RGSFGYAAUTVSQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

remainder C5 and C6 hydrocarbons

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenwei Liu et al.
Physical chemistry chemical physics : PCCP, 17(30), 20021-20029 (2015-07-15)
Clathrate hydrate particle agglomeration is often considered to be one of the key limiting factors in plug formation. The hydrate particle-water interaction can play a critical role in describing hydrate agglomeration, yet is severely underexplored. Therefore, this work investigates the
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Shane Avison et al.
Journal of agricultural and food chemistry, 63(28), 6313-6318 (2015-06-23)
A new methodology is presented to measure water-air partition coefficients (Henry's constants) of volatiles, using APCI-MS. Significant advantages over other Henry's constant determination methods include the short measurement and sample preparation time and the possibility for simultaneous measurement of multiple
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service