Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278092

Sigma-Aldrich

Activated charcoal

DARCO®, wet powder, −100 mesh

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12000000

vapor pressure

<0.1 mmHg ( 20 °C)

form

wet powder

autoignition temp.

842 °F

contains

<30% water

manufacturer/tradename

DARCO®

resistivity

1375 μΩ-cm, 20°C (graphite)

particle size

−100 mesh

pore size

~1.5 cm3/g pore volume (dry basis)

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Suitable for decolorizing and purifying pharmaceuticals. Its excellent adsorptive capacity is especially effective in adsorbing higher molecular weight molecules, resulting in colorless liquids or white crystals.

Legal Information

DARCO is a registered trademark of Cabot Norit Americas, Inc.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service