25116
Sodium citrate tribasic dihydrate
puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E331, 99-100.5% (calc. to the dried substance)
Synonym(s):
Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate
About This Item
Recommended Products
grade
puriss.
Quality Level
assay
99-100.5% (calc. to the dried substance)
quality
meets analytical specification of Ph. Eur., BP, USP, FCC, E331
impurities
Residual solvent, in accordance
acidity or alkalinity, complies
≤0.0005% heavy metals (as Pb)
≤0.04% free alkali (as NaOH)
≤0.06% free acid (as C6H8O7)
11.0-13.0 water (Karl Fischer)
loss
11-13% loss on drying, 180 °C, 18 h
pH
7.5-9.0 (20 °C, 5%)
mp
>300 °C (lit.)
anion traces
chloride (Cl-): ≤30 mg/kg
oxalate (as oxalic acid): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg
tartrate (C4H4O62-): in accordance
cation traces
As: ≤1 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤10 mg/kg
suitability
alkaline for reaction against litmus
complies for appearance of solution
complies for reaction against H2SO4
SMILES string
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
InChI
1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3
InChI key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service