Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25006

Sigma-Aldrich

Ammonium acetate

puriss., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein/REAXYS Number:
4186741
EC Number:
MDL number:
UNSPSC Code:
12161700

grade

puriss.

assay

≥98%

form

solid

impurities

≤0.0005% heavy metals (as Pb)

ign. residue

≤0.02% (as SO4)

mp

110-112 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤5 mg/kg

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-François Daudelin et al.
Veterinary research, 42, 69-69 (2011-05-25)
This study evaluated the effect of the probiotics Pediococcus acidilactici and Saccharomyces cerevisiae boulardii on the intestinal colonization of O149 enterotoxigenic Escherichia coli harbouring the F4 (K88) fimbriae (ETEC F4) and on the expression of ileal cytokines in weaned pigs.
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype
Suzanne L Parker et al.
Journal of pharmaceutical and biomedical analysis, 105, 39-45 (2014-12-22)
A high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method, using hydrophilic interaction liquid chromatography (HILIC) chromatography for the analysis of fosfomycin in human plasma and urine, has been developed and validated. The plasma method uses a simple protein precipitation using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service