Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

244279

Sigma-Aldrich

Hydrofluoric acid

ACS reagent, 48%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

assay

48%
48.0-51.0% (ACS specification)

form

liquid

impurities

≤0.01% fluorosilicic acid

ign. residue

≤5 ppm

density

1.15 g/mL at 25 °C (lit.)

anion traces

SO42- and SO32-: ≤5 ppm
chloride (Cl-): ≤5 ppm
phosphate (PO43-): ≤1 ppm

cation traces

As: ≤0.005 ppm
Cu: ≤0.1 ppm
Fe: ≤1 ppm
heavy metals (as Pb): ≤0.5 ppm

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian Ford et al.
International journal of cardiology, 184, 163-169 (2015-02-24)
We identified easily obtained baseline characteristics associated with outcomes in patients with chronic heart failure (HF) and elevated heart rate (HR) receiving contemporary guideline-recommended therapy in the SHIFT trial, and used them to develop a prognostic model. We selected the
Yvonne C J Wientjes et al.
Genetics, selection, evolution : GSE, 47, 42-42 (2015-05-09)
Although simulation studies show that combining multiple breeds in one reference population increases accuracy of genomic prediction, this is not always confirmed in empirical studies. This discrepancy might be due to the assumptions on quantitative trait loci (QTL) properties applied
Wenlin Yvonne Lin et al.
Waste management (New York, N.Y.), 41, 75-84 (2015-04-22)
Accelerated carbonation has been studied as a treatment method for MSW IBA, and the main advantage is that it can shorten the treatment duration from months to days, compared to natural weathering. This study investigated the effect of accelerated carbonation
Faiez Zannad et al.
European journal of heart failure, 17(7), 735-742 (2015-04-29)
Thrombin is a critical element of crosstalk between pathways contributing to worsening of established heart failure (HF). The aim of this study is to explore the efficacy and safety of rivaroxaban 2.5 mg bid compared with placebo (with standard care) after
Nicolas Vodovar et al.
JACC. Heart failure, 3(8), 629-636 (2015-08-08)
This study sought to hypothesize that elevated B-type natriuretic peptide (BNP) could act as an endogenous neprilysin inhibitor. A hallmark of acute decompensated heart failure (ADHF) is the overproduction of natriuretic peptides (NPs) by stretched cardiomyocytes. Various strategies have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service