Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

242799

Sigma-Aldrich

1-Methyl-2-pyrrolidinone

spectrophotometric grade, ≥99%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein/REAXYS Number:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

assay

≥99%

autoignition temp.

518 °F

expl. lim.

9.5 %

impurities

≤0.05% water (Karl Fischer)

refractive index

n20/D 1.47 (lit.)

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 285 nm Amax: 1.00
λ: 300 nm Amax: 0.50
λ: 325 nm Amax: 0.10
λ: 350-400 nm Amax: 0.01

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

195.8 °F - Pensky-Martens closed cup

flash_point_c

91 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Ping Neo et al.
Journal of agricultural and food chemistry, 62(22), 5163-5172 (2014-05-16)
Effects of heat treatment on structure and physicochemical properties of zein (Ze) and gallic acid loaded zein (Ze-GA) electrospun fiber mats were investigated. The electrospun fiber mats displayed different surface and physicochemical properties after being heat-cured at 150 °C for
B A Jönsson et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(2), 351-357 (1997-07-04)
A method for simultaneous determination of 5-hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide in urine is described. These compounds are metabolites of N-methyl-2-pyrrolidone, a powerful and widely used organic solvent. 5-Hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide were purified from urine by adsorption to a C8 solid-phase extraction
Laura Vogelaar et al.
Small (Weinheim an der Bergstrasse, Germany), 1(6), 645-655 (2006-12-29)
Phase separation micromolding (PSmicroM) is a versatile microfabrication technique that can be used to structure a very broad range of polymers, including block copolymers and biodegradable and conductive polymers without the need for clean-room facilities. By incorporating a subsequent process
Chun-Wa Chung et al.
Journal of medicinal chemistry, 55(2), 576-586 (2011-12-06)
Bromodomain-containing proteins are key epigenetic regulators of gene transcription and readers of the histone code. However, the therapeutic benefits of modulating this target class are largely unexplored due to the lack of suitable chemical probes. This article describes the generation
Ajit Dhananjay Jagtap et al.
European journal of medicinal chemistry, 85, 268-288 (2014-08-05)
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50 = 0.4 nM and 0.5 nM, respectively).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service