242179
Silica gel
high-purity grade, pore size 40 Å, 35-70 mesh particle size
About This Item
Recommended Products
grade
high-purity grade
form
powder
manufacturer/tradename
EMD Millipore® 10181
technique(s)
LPLC: suitable
matrix
Silica
matrix active group
silica
particle size
200-500 μm
35-70 mesh
pore size
~0.68 cm3/g pore volume
40 Å
bp
2230 °C
mp
>1600 °C
separation technique
hydrophilic interaction (HILIC)
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction.[1]
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service