236039
Fat Brown RR
Dye content 95 %
Synonym(s):
Solvent Brown 1
About This Item
composition
Dye content, 95%
mp
147-158 °C (lit.)
λmax
451 nm
SMILES string
Nc1ccc(N=Nc2cccc3ccccc23)c(N)c1
InChI
1S/C16H14N4/c17-12-8-9-16(14(18)10-12)20-19-15-7-3-5-11-4-1-2-6-13(11)15/h1-10H,17-18H2
InChI key
AHWMWMNEYBHQNL-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service