Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217093

Sigma-Aldrich

Disperse Orange 11

Dye content 95 %

Synonym(s):

1-Amino-2-methylanthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
Colour Index Number:
60700
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 95%

mp

204-206 °C (lit.)

λmax

486 nm

SMILES string

Cc1ccc2C(=O)c3ccccc3C(=O)c2c1N

InChI

1S/C15H11NO2/c1-8-6-7-11-12(13(8)16)15(18)10-5-3-2-4-9(10)14(11)17/h2-7H,16H2,1H3

InChI key

ZLCUIOWQYBYEBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service