Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

215384

Sigma-Aldrich

Potassium fluoride

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein/REAXYS Number:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
98%
grade:
reagent grade

grade

reagent grade

assay

98%

mp

858 °C (lit.)

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natural phosphate doped with potassium fluoride and modified with sodium nitrate: Efficient catalysts for the Knoevenagel condensation.
Sebti S, et al.
Tetrahedron Letters, 43(10), 1813-1815 (2002)
Selective desilylation of tert-butyldimethylsilyl ethers of phenols using potassium fluoride-alumina and ultrasound.
Schmittling EA and Sawyer JS.
Tetrahedron Letters, 32(49), 7207-7210 (1991)
Potassium fluoride on celite. A versatile reagent for C-, N-, O-, and S-alkylations.
Ando T and Yamawaki J.
Chemistry Letters (Jpn), 1, 45-46 (1979)
Characterization of the actual catalytic agent in potassium fluoride on activated alumina systems.
Weinstock LM, et al.
Tetrahedron Letters, 27(33), 3845-3848 (1986)
Mingming Fan et al.
Bioresource technology, 104, 447-450 (2011-12-16)
Transesterification of soybean oil with methanol was carried out in the presence of CaO-MgO and KF-modified CaO-MgO catalysts at atmospheric pressure. While the methyl ester yield for the CaO-MgO catalyst with a ratio of 8:2 (CaO:MgO) was 63.6%, it was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service