Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

210390

Sigma-Aldrich

Acid Red 97

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H20N4Na2O8S2
CAS Number:
Molecular Weight:
698.63
Colour Index Number:
22890
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 40%

λmax

498 nm

SMILES string

[Na+].[Na+].Oc1ccc2ccccc2c1\N=N\c3ccc(-c4ccc(cc4S([O-])(=O)=O)\N=N\c5c(O)ccc6ccccc56)c(c3)S([O-])(=O)=O

InChI

1S/C32H22N4O8S2.2Na/c37-27-15-9-19-5-1-3-7-23(19)31(27)35-33-21-11-13-25(29(17-21)45(39,40)41)26-14-12-22(18-30(26)46(42,43)44)34-36-32-24-8-4-2-6-20(24)10-16-28(32)38;;/h1-18,37-38H,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2/b35-33+,36-34+;;

InChI key

JCEBMROGCIEFRX-UUIOKUIJSA-L

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service