Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

198137

Sigma-Aldrich

Crocein Orange G

Dye content 70 %

Synonym(s):

1-Phenylazo-2-naphthol-6-sulfonic acid sodium salt, Acid Orange 12, Ponceau 4 GB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N2NaO4S
CAS Number:
Molecular Weight:
350.32
Colour Index Number:
15970
Beilstein/REAXYS Number:
4117392
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 70%

λmax

482 nm

SMILES string

[Na+].Oc1ccc2cc(ccc2c1\N=N\c3ccccc3)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service