Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

192325

Sigma-Aldrich

4-Methyl-3-nitrobenzyl chloride

97%

Synonym(s):

3-nitro-4-methylbenzyl chloride, 4-(Chloromethyl)-1-methyl-2-nitrobenzene, 4-(Chloromethyl)-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2Cl
CAS Number:
Molecular Weight:
185.61
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

assay

97%

form

crystalline

bp

87-88 °C/0.05 mmHg (lit.)

mp

48-50 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(CCl)cc1[N+]([O-])=O

InChI

1S/C8H8ClNO2/c1-6-2-3-7(5-9)4-8(6)10(11)12/h2-4H,5H2,1H3

InChI key

DLTKEOYTIZIUKD-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service