Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18710

Sigma-Aldrich

Hydrobromic acid

puriss. p.a., ACS reagent, dist., ≥48% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
CAS Number:
Molecular Weight:
80.91
Beilstein/REAXYS Number:
3587158
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

vapor density

2.8 (vs air)

vapor pressure

320 psi ( 21.1 °C)
8 mmHg ( 25 °C)

description

48% aqueous HBr

assay

≥48% (T)

quality

dist.

impurities

≤20 mg/kg total iodine

ign. residue

≤0.002% (as SO4)

refractive index

n20/D 1.438

density

1.49 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate, sulfite (as SO42-): ≤30 mg/kg

cation traces

Ag: ≤0.05 mg/kg
Al: ≤0.1 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.1 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.2 mg/kg
K: ≤0.2 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.01 mg/kg
Se: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Reagent for the decomposition of metals, oxides, carbonates and magnetite[1][2]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z. Sulcek et al.
Methods of Decomposition in Inorganic Analysis (1989)
R. Bock
Aufschlussmethoden der anorg. & org. Chemie (1972)
Po-Yu Tsai et al.
Physical chemistry chemical physics : PCCP, 13(4), 1419-1423 (2010-11-27)
The orientation dependence of Br-atom formation in the reaction of the oriented OH radical with the HBr molecule using the hexapole electrostatic field was studied. Experimental results for the orientation dependence in the reaction were analyzed using a Legendre polynomial
Małgorzata Olejniczak et al.
The Journal of chemical physics, 136(1), 014108-014108 (2012-01-14)
We report the implementation of nuclear magnetic resonance (NMR) shielding tensors within the four-component relativistic Kohn-Sham density functional theory including non-collinear spin magnetization and employing London atomic orbitals to ensure gauge origin independent results, together with a new and efficient
Ahmed Hasbi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(11), 4806-4820 (2014-07-27)
Although the dopamine D1-D2 receptor heteromer has emerging physiological relevance and a postulated role in different neuropsychiatric disorders, such as drug addiction, depression, and schizophrenia, there is a need for pharmacological tools that selectively target such receptor complexes in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service