17741
Diethylene glycol diglycidyl ether
technical, ~50% (GC)
Synonym(s):
Diglycidyl diglycol
About This Item
Recommended Products
grade
technical
concentration
~50% (GC)
refractive index
n20/D 1.467
density
1.167 g/mL at 20 °C (lit.)
SMILES string
C(COCC1CO1)OCCOCC2CO2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service