Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15760

Sigma-Aldrich

D-(−)-Fructose

meets analytical specification of Ph. Eur., BP

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

optical activity

[α]20/D −93.5 to −91.0°, c = 10% in H2O

quality

meets analytical specification of Ph. Eur., BP

impurities

5-hydroxymethylfurfurol and related substances, complies
acidity or alcalinity, complies
foreign sugar, complies
residual solvents, complies
≤0.5% water (Karl Fischer)

ign. residue

≤0.1% (as SO4)

mp

119-122 °C (dec.) (lit.)

cation traces

Ba:, in accordance
Pb: ≤0.5 mg/kg

suitability

in accordance for appearance of solution

application(s)

pharmaceutical (small molecule)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(-)-Fructose was used as a test compound in NMR study for examining the mechanism of dehydration of D-fructose to 5-hydroxymethylfurfural in DMSO at 150°C.[1]

Biochem/physiol Actions

D-(-)-Fructose enhances mood and causes gastrointestinal disturbances in fructose malabsorbers.[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ananda S Amarasekara et al.
Carbohydrate research, 343(18), 3021-3024 (2008-10-03)
The anomeric composition of d-fructose in dimethyl sulfoxide changes when the solution is heated from room temperature to 150 degrees C, with a small increase in the alpha-furanose form at the expense of the beta-pyranose tautomer. Additionally, a small amount
M Ledochowski et al.
Scandinavian journal of gastroenterology, 35(10), 1048-1052 (2000-12-01)
Fructose malabsorption is characterized by the inability to absorb fructose efficiently. As a consequence fructose reaches the colon where it is broken down by bacteria to short fatty acids, CO2 and H2. Bloating, cramps, osmotic diarrhea and other symptoms of
B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
K Shiraga et al.
Carbohydrate research, 406, 46-54 (2015-02-07)
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service