Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15329

Sigma-Aldrich

Cyclohexane

puriss., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein/REAXYS Number:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

grade

puriss.

assay

≥99.5% (GC)

impurities

≤0.0015% non-volatile matter
≤0.02% water (Karl Fischer)

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

Inchi Key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane participates as starting reagent in the photonitrosylation process (PNC process) and affords ε-caprolactam. Its oxidation by employing various oxidants such as hydrogen peroxide, tert-butyl hydroperoxide and molecular oxygen has been reported.

Application

Cyclohexane has been used to prepare the 1.3M solution of sec-butyllithium. It may be used as starting reagent for the synthesis of adipic acid.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pertains only in Germany: unversteuert

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched nanomolecules: Regular polystyrene dendrigrafts.
Muchtar Z, et al.
Macromolecules, 34(22), 7595-7600 (2001)
Cyclohexane oxidation continues to be a challenge.
Schuchardt ULF, et al.
Applied Catalysis A: General, 211(1), 1-17 (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 168-168 (1994)
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Robert Duran et al.
Environmental science and pollution research international, 22(20), 15215-15229 (2015-02-26)
The exploration of marine Actinobacteria has as major challenge to answer basic questions of microbial ecology that, in turn, will provide useful information to exploit Actinobacteria metabolisms in biotechnological processes. The ecological functions performed by Actinobacteria in marine sediments are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service