Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13412

Sigma-Aldrich

Sodium azide

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
EC Number:
MDL number:
UNSPSC Code:
12352000
eCl@ss:
38060110

assay

≥99%

impurities

≤0.001% heavy metals (as Pb)
≤0.05% insoluble in water
≤0.2% free alkali (as NaOH)

loss

≤0.2% loss on drying (on P2O5,20 mbar)

application(s)

agriculture
environmental

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

suggested gloves for splash protection

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

target_organs

Brain

supp_hazards

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyi Huang et al.
Lasers in surgery and medicine, 44(6), 490-499 (2012-07-05)
Antimicrobial photodynamic therapy (APDT) employs a non-toxic photosensitizer (PS) and visible light, which in the presence of oxygen produce reactive oxygen species (ROS), such as singlet oxygen ((1) O(2), produced via Type II mechanism) and hydroxyl radical (HO(.), produced via
Gilles Lambert et al.
Journal of the American College of Cardiology, 63(22), 2365-2373 (2014-03-19)
Do elevated proprotein convertase subtilisin/kexin type 9 (PCSK9) levels constitute an even greater risk for patients who already have reduced low-density lipoprotein receptor (LDLR) levels, such as those with heterozygous familial hypercholesterolemia (HeFH)? As a circulating inhibitor of LDLR, PCSK9
Andrew J Kassianos et al.
Kidney international, 87(6), 1153-1163 (2015-01-15)
Chemokines play pivotal roles in tissue recruitment and retention of leukocytes, with CX3CR1 recently identified as a chemokine receptor that selectively targets mouse kidney dendritic cells (DCs). We have previously demonstrated increased tubulointerstitial recruitment of human transforming growth factor-β (TGF-β)-producing
Angela S W Tjon et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5625-5634 (2014-05-09)
High-dose i.v. Ig (IVIg) is a prominent immunomodulatory therapy for various autoimmune and inflammatory diseases. Recent mice studies suggest that IVIg inhibits myeloid cell function by inducing a cascade of IL-33-Th2 cytokine production causing upregulation of the inhibitory FcγRIIb, as
Luisa M Villar et al.
Annals of neurology, 76(2), 231-240 (2014-06-10)
To identify a biomarker distinguishing patients who, despite a primary progressive multiple sclerosis (PPMS) clinical course, may nonetheless benefit from immune therapy. The presence or absence of both immunoglobulin (Ig) G and IgM oligoclonal bands (OCB) was blindly examined in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service