Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13301

Sigma-Aldrich

Ammonium molybdate tetrahydrate

puriss., ≥99%

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss.

assay

≥99%

impurities

≤0.002% heavy metals (as Pb)

density

2.498 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
phosphate, arsenate, silicate (as SiO2): ≤10 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Fe: ≤20 mg/kg

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Cornish et al.
Journal of the American Veterinary Medical Association, 231(4), 586-589 (2007-08-19)
A closed herd of 400 mixed-breed dairy goats was examined because of a decrease in milk production and increase in mortality rate. Nine animals had died within a 1-month period. Clinical signs were evident only in lactating goats and included
Shifeng Cao et al.
International journal of food microbiology, 141(3), 173-176 (2010-07-08)
The potential enhancement of Pichia membranifaciens by ammonium molybdate (NH(4)Mo) to control blue mould caused by Penicillium expansum on peach fruit was investigated. Combining P. membranifaciens at 1x10(8) cell/ml with 1 mM NH(4)Mo provided a more effective control of blue
Pradeep Verma et al.
Bioresource technology, 102(4), 3941-3945 (2010-12-24)
Pretreatments for enzymatic saccharification are crucial for the establishment of lignocellulosic biorefineries. In this study, we focused on ammonium ions and peroxometal complexes as potential delignifying agents. We first examined the pretreatment of beech wood with nine different ammonium salts
Laura Borgnino et al.
Talanta, 85(3), 1310-1316 (2011-08-03)
This paper proposes a new automated spectrophotometric method for the simultaneous determination of phosphate and arsenate without pre-treatment, which is faster, simpler, less expensive and hazardous than other well-known methods used with water samples. Such method is based on the
Sudarat Yenjai et al.
Biochemical and biophysical research communications, 419(1), 126-129 (2012-02-15)
In this study, the cleavage of protein by molybdenum cluster is reported for the first time. The protein target used is porcine pepsin. The data presented in this study show that pepsin is cleaved to at least three fragments with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service