Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12659

Sigma-Aldrich

Potassium sulfate

puriss., ≥99% (calc. to the dried substance), powder (fine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

puriss.

assay

≥99% (calc. to the dried substance)

form

powder (fine)

impurities

≤0.001% heavy metals (as Pb)
≤0.01% free acid (as H2SO4)
≤0.03% free alkali (as KOH)

loss

≤0.5% loss on drying, 105 °C, 4 h

granulation

<0.3 mm

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

As: ≤5 mg/kg
Fe: ≤50 mg/kg

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service