Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

121266

Sigma-Aldrich

Acriflavine hydrochloride

Dye content 90 %

Synonym(s):

3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein/REAXYS Number:
6265525
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 90%

mp

260 °C (dec.) (lit.)

λmax

465 nm

SMILES string

[Cl-].Cl[H].Cl[H].Nc1ccc2cc3ccc(N)cc3nc2c1.C[n+]4c5cc(N)ccc5cc6ccc(N)cc46

InChI

1S/C14H13N3.C13H11N3.3ClH/c1-17-13-7-11(15)4-2-9(13)6-10-3-5-12(16)8-14(10)17;14-10-3-1-8-5-9-2-4-11(15)7-13(9)16-12(8)6-10;;;/h2-8H,1H3,(H3,15,16);1-7H,14-15H2;3*1H

Inchi Key

MKLTXAHQKDVBLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A mixture of 3,6-diamino-10-methylacridinium (acriflavin) and 3,6-diaminoacridine (proflavin)

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RNA synthesis by isolated mammalian mitochondria and nuclei: effects of ethidium bromide and acriflavin..
R R Meyer et al.
Archives of biochemistry and biophysics, 148(2), 425-430 (1972-02-01)
Biosynthesis of yeast mitochondria. Drug effects on the petite negative yeast kluyveromyces lactis.
A A Luha et al.
Biochemical and biophysical research communications, 44(2), 396-402 (1971-07-16)
Euflavin and ethidium bromide; inhibitors of mitochondriogenesis in regenerating rat liver.
H De Vries et al.
FEBS letters, 7(4), 347-350 (1970-05-01)
Shizuka Nakayama et al.
Chemical communications (Cambridge, England), 47(16), 4766-4768 (2011-03-15)
The ubiquitous bacterial biofilm regulator, c-di-GMP can form G-quadruplexes at physiological conditions in the presence of some aromatic compounds, such as acriflavine and proflavine. The fluorescence of these compounds is quenched upon c-di-GMP binding and some of the formed c-di-GMP
Ho To et al.
The Journal of veterinary medical science, 74(7), 949-953 (2012-03-27)
The objective of the present study was to characterize Erysipelothrix sp. strains from recent erysipelas outbreaks in Japan. Eighty-three (100%) strains were identified as E. rhusiopathiae, based on serotyping and spaA PCR. Fifty (60.3%), 5 (6.0%), and 28 (33.7%) strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service